Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: efl-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129186.3 | NP_001122658.1 | 2 - 3 | 463 | 80 | |
NM_001129188.4 | NP_001122660.1 | 2 - 3 | 463 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |