Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tps-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064634.6 | NP_497035.3 | 5 - 6 | 1600 | 76 | |
GenBank mRNA | AJ811574.1 | - | 5 - 6 | 1622 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |