Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267510.3 | NP_001254439.1 | 10 - 11 | 2689 | 105 | |
NM_001267513.3 | NP_001254442.1 | 11 - 12 | 3557 | 105 | ||
NM_001267514.4 | NP_001254443.1 | 11 - 12 | 3466 | 105 | ||
NM_001267515.3 | NP_001254444.1 | 11 - 12 | 3466 | 105 | ||
NM_001267516.3 | NP_001254445.1 | 11 - 12 | 3466 | 105 | ||
NM_001267517.3 | NP_001254446.1 | 11 - 12 | 3466 | 105 | ||
NM_001350388.3 | NP_001337294.1 | 20 | 5833 | 105 | ||
NM_064643.10 | NP_497044.3 | 11 - 12 | 3466 | 105 | ||
NM_064644.6 | NP_497045.1 | 11 - 12 | 3556 | 105 | ||
NM_064645.6 | NP_497046.1 | 11 - 12 | 3557 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |