Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027640.5 | NP_001022811.1 | 6 - 7 | 1104 | 74 | |
NM_001381690.1 | NP_001367960.1 | 9 - 10 | 1405 | 74 | ||
NM_001384006.1 | 5 - 6 | 1521 | 74 | |||
NM_064789.5 | NP_497190.1 | 11 - 12 | 1907 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |