Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceeh-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064867.6 | NP_497268.1 | 3 - 4 | 784 | 78 | |
GenBank mRNA | EU151493.1 | - | 3 - 4 | 790 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |