Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064876.9 | NP_497277.2 | 1 - 2 | 158 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |