Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027761.7 | NP_001022932.1 | 2 - 3 | 226 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |