Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129227.4 | NP_001122699.2 | 16 - 17 | 3837 | 87 | |
NM_001267920.3 | NP_001254849.1 | 16 - 17 | 3837 | 87 | ||
NM_001373851.2 | NP_001360184.1 | 3 | 972 | 87 | ||
NM_065077.6 | NP_497478.2 | 16 - 17 | 3846 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |