Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpt-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065097.5 | NP_497498.2 | 6 - 7 | 1935 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |