Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026047.5 | NP_001021218.1 | 3 - 4 | 433 | 68 | |
NM_001381807.1 | NP_001366876.1 | 3 - 4 | 421 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |