Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbb-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065405.8 | NP_497806.1 | 1 - 2 | 170 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |