Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wht-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001322779.3 | NP_001309589.1 | 1 | 91 | 75 | |
NM_065424.3 | NP_497825.2 | 1 - 2 | 138 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |