Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gly-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027473.4 | NP_001022644.1 | 3 - 4 | 519 | 74 | |
NM_001027475.3 | NP_001022646.1 | 2 - 3 | 243 | 74 | ||
NM_001384070.1 | 2 - 3 | 243 | 74 | |||
GenBank mRNA | AF031838.1 | - | 2 - 3 | 250 | 74 | |
AF031839.1 | - | 2 - 3 | 250 | 74 | ||
AF031840.1 | - | 2 - 3 | 250 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |