Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sel-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377920.3 | NP_001364559.1 | 30 - 31 | 7093 | 96 | |
NM_065538.6 | NP_497939.2 | 29 - 30 | 6958 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |