Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065613.9 | NP_498014.2 | 7 - 8 | 2242 | 151 |
Gene Symbol: C35D10.17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381846.1 | NP_001367631.1 | 5 - 6 | 1942 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |