Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nca-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136363.4 | NP_001129835.1 | 17 - 18 | 4210 | 80 | |
NM_065653.6 | NP_498054.2 | 18 - 19 | 4366 | 80 | ||
GenBank mRNA | AY555272.2 | - | 18 - 19 | 4366 | 80 | |
DQ917241.1 | - | 17 - 18 | 4248 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |