Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gop-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_065717.6 | NP_498118.1 | 1 - 2 | 157 | 72 | |
GenBank mRNA | AY052770.1 | - | 1 - 2 | 157 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |