Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026014.4 | NP_001021185.2 | 8 - 9 | 831 | 74 | |
NM_001026015.3 | NP_001021186.1 | 4 - 5 | 629 | 74 | ||
NM_001129205.3 | NP_001122677.1 | 6 - 7 | 654 | 74 | ||
GenBank mRNA | EU239658.1 | - | 8 - 9 | 826 | 74 | |
EU239659.1 | - | 4 - 5 | 565 | 74 | ||
EU239660.1 | - | 6 - 7 | 643 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |