Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyk-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027821.6 | NP_001022992.1 | 10 - 11 | 2398 | 77 | |
NM_065910.7 | NP_498311.2 | 10 - 11 | 2383 | 77 | ||
GenBank mRNA | AF469173.1 | - | 10 - 11 | 2336 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |