Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sma-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066092.4 | NP_498493.1 | 9 - 10 | 960 | 92 | |
GenBank mRNA | U34902.1 | - | 9 - 10 | 961 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |