Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osm-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330924.2 | NP_001317784.1 | 5 - 6 | 657 | 83 | |
NM_066197.7 | NP_498598.1 | 4 - 5 | 776 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |