Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379826.1 | NP_001367746.1 | 7 - 8 | 1039 | 116 | |
NM_001382935.1 | 7 - 8 | 1043 | 116 | |||
NM_066420.6 | NP_498821.2 | 5 - 6 | 673 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |