Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rfp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027528.5 | NP_001022699.1 | 4 - 5 | 921 | 78 | |
NM_001027529.6 | NP_001022700.1 | 4 - 5 | 924 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |