Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136361.5 | NP_001129833.1 | 11 - 12 | 2067 | 97 | |
NM_001306725.3 | NP_001293654.1 | 3 - 4 | 577 | 97 | ||
NM_001306727.4 | NP_001293656.1 | 5 - 6 | 1037 | 97 | ||
NM_001306728.4 | NP_001293657.1 | 11 - 12 | 2077 | 97 | ||
NM_066476.7 | NP_498877.4 | 11 - 12 | 1978 | 97 | ||
GenBank mRNA | EU752496.1 | - | 11 - 12 | 2067 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |