Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnj-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066500.6 | NP_498901.1 | 5 - 6 | 743 | 69 | |
NM_066501.4 | NP_498902.1 | 5 - 6 | 743 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |