Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066512.6 | NP_498913.2 | 2 - 3 | 311 | 70 | |
GenBank mRNA | U01183.1 | - | 3 - 4 | 344 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |