Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066642.6 | NP_499043.1 | 6 - 7 | 953 | 88 | |
GenBank mRNA | AF079899.1 | - | 6 - 7 | 961 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |