Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crn-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382961.1 | 4 - 5 | 505 | 123 | ||
NM_066660.5 | NP_499061.2 | 4 - 5 | 540 | 123 | ||
GenBank mRNA | AY303580.1 | - | 4 - 5 | 505 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |