Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026054.4 | NP_001021225.1 | 7 - 8 | 2622 | 86 | |
NM_001268083.3 | NP_001255012.1 | 7 - 8 | 2681 | 86 | ||
NM_001379863.1 | NP_001366984.1 | 6 - 7 | 2519 | 86 | ||
NM_066714.4 | NP_499115.2 | 7 - 8 | 2630 | 86 | ||
GenBank mRNA | AF049142.1 | - | 7 - 8 | 2682 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |