Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncx-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066745.3 | NP_499146.2 | 1 - 2 | 208 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |