Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: T05G5.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268094.3 | NP_001255023.1 | 3 - 4 | 828 | 73 |
Gene Symbol: ech-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379869.2 | NP_001366674.1 | 3 - 4 | 867 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |