Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rde-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066864.5 | NP_499265.1 | 3 - 4 | 733 | 121 | |
GenBank mRNA | AY071926.1 | - | 3 - 4 | 731 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |