Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027611.3 | NP_001022782.1 | 14 - 15 | 1770 | 122 | |
NM_001027612.4 | NP_001022783.1 | 6 - 7 | 917 | 122 | ||
NM_001027613.3 | NP_001022784.1 | 6 - 7 | 917 | 122 | ||
NM_001027614.4 | NP_001022785.1 | 6 - 7 | 836 | 122 | ||
NM_001379901.2 | NP_001368252.1 | 6 - 7 | 836 | 122 | ||
NM_001379902.1 | NP_001368254.1 | 2 - 3 | 207 | 122 | ||
NM_001379904.1 | NP_001367142.1 | 6 - 7 | 917 | 122 | ||
NM_001382981.1 | 10 - 11 | 1732 | 122 | |||
GenBank mRNA | AF151641.1 | - | 6 - 7 | 843 | 122 | |
AF151642.1 | - | 6 - 7 | 843 | 122 | ||
AF151643.1 | - | 6 - 7 | 843 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |