Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnt-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027666.5 | NP_001022837.1 | 3 - 4 | 295 | 70 | |
NM_001382988.1 | 3 - 4 | 292 | 70 | |||
NM_001382989.1 | 3 - 4 | 271 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |