Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oxi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066991.5 | NP_499392.1 | 10 - 11 | 2301 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |