Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027713.5 | NP_001022884.1 | 12 - 13 | 1840 | 75 | |
NM_001372661.1 | NP_001359517.1 | 12 - 13 | 1806 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |