Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067367.8 | NP_499768.2 | 4 - 5 | 636 | 80 |
Gene Symbol: ttr-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383023.1 | 6 - 7 | 1245 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Caenorhabditis elegans Probe spans exons |