Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cua-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268273.3 | NP_001255202.1 | 8 - 9 | 1380 | 70 | |
NM_001268274.3 | NP_001255203.1 | 5 - 6 | 881 | 70 | ||
GenBank mRNA | D83665.1 | - | 7 - 8 | 1316 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |