Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdk-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067382.8 | NP_499783.1 | 5 - 6 | 883 | 86 | |
GenBank mRNA | AF129111.1 | - | 5 - 6 | 788 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |