Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aqp-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067420.6 | NP_499821.2 | 4 - 5 | 628 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |