Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027836.5 | NP_001023007.1 | 7 - 8 | 2401 | 60 | |
NM_001027837.3 | NP_001023008.1 | 7 - 8 | 2241 | 60 | ||
NM_001379988.1 | NP_001367020.1 | 7 - 8 | 2119 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |