Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: toca-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067437.6 | NP_499838.2 | 6 - 7 | 804 | 64 | |
NM_067438.5 | NP_499839.2 | 6 - 7 | 852 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |