Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307237.2 | NP_001294166.1 | 5 - 6 | 885 | 76 | |
NM_001307238.2 | NP_001294167.1 | 5 - 6 | 1036 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |