Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ora-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068933.7 | NP_501334.1 | 1 - 2 | 112 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |