Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sax-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038306.5 | NP_001033395.1 | 4 - 5 | 654 | 108 | |
NM_001038307.5 | NP_001033396.1 | 4 - 5 | 664 | 108 | ||
GenBank mRNA | AB206669.1 | - | 4 - 5 | 591 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |