Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabs-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307243.2 | NP_001294172.1 | 3 - 4 | 497 | 86 | |
NM_068951.4 | NP_501352.1 | 3 - 4 | 499 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |