Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hgrs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068974.8 | NP_501375.2 | 7 - 8 | 2170 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |