Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306928.3 | NP_001293857.1 | 18 - 19 | 5220 | 77 | |
NM_001306929.3 | NP_001293858.1 | 18 - 19 | 5245 | 77 | ||
NM_001380409.1 | NP_001368188.1 | 19 - 20 | 5276 | 77 | ||
NM_001380410.1 | NP_001366749.1 | 19 - 20 | 5276 | 77 | ||
NM_069252.7 | NP_501653.1 | 18 - 19 | 5236 | 77 | ||
GenBank mRNA | AY396716.1 | - | 19 - 20 | 5276 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |