Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307073.3 | NP_001294002.1 | 4 - 5 | 532 | 107 | |
NM_069323.3 | NP_501724.2 | 4 - 5 | 560 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |