Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069327.3 | NP_501728.2 | 4 - 5 | 493 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |